siRNA Details
Gene Symbol: Mllt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106876.1 | NP_001100346.1 | 12 | 1775 | ||
XM_006244307.2 | XP_006244369.1 | 11 | 1645 |

More Information
Other Species Targeted

Gene Ontology Categories:
