siRNA Details
Gene Symbol: Rfx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106877.1 | NP_001100347.1 | 13 | 1655 | ||
XM_006244313.2 | XP_006244375.1 | 14 | 1745 | |||
GenBank mRNA | BC166527.1 | - | 13 | 1572 | 7135787 |
