siRNA Details
Gene Symbol: Dus3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034923.1 | NP_001030095.1 | 6 | 1143 | ||
XM_006244314.2 | XP_006244376.1 | 6 | 1298 | |||
GenBank mRNA | BC105780.1 | - | 6 | 1143 | 7932299 |
