siRNA Details
Gene Symbol: Dpp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244369.2 | XP_006244431.1 | 3 | 958 | ||
XM_008766803.1 | XP_008765025.1 | 3 | 329 | |||
XM_008766804.1 | XP_008765026.1 | 2 | 1139 | |||
XM_217309.8 | XP_217309.6 | 3 | 958 |
