siRNA Details
Gene Symbol: Tnfsf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191803.1 | NP_001178732.1 | 4 | 330 | ||
XM_008766755.1 | XP_008764977.1 | 4 | 385 |

More Information
Other Species Targeted

Gene Ontology Categories:
