siRNA Details
Gene Symbol: St6gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100888.1 | NP_001094358.1 | 4 | 1234 | ||
XM_006244260.2 | XP_006244322.1 | 3 | 1439 | |||
XM_006244264.2 | XP_006244326.1 | 3 | 1302 | |||
XM_006244265.2 | XP_006244327.1 | 3 | 1606 | |||
XM_008766756.1 | XP_008764978.1 | 3 | 3409 | |||
XM_008766757.1 | XP_008764979.1 | 3 | 1692 | |||
XM_008766758.1 | XP_008764980.1 | 3 | 3409 | |||
GenBank mRNA | AJ627626.1 | - | 2 | 1012 |

More Information
Other Species Targeted

Gene Ontology Categories:
