siRNA Details
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244415.2 | XP_006244477.1 | 12 | 2012 | ||
XM_006244416.2 | XP_006244478.1 | 11 | 1953 | |||
XM_008766840.1 | XP_008765062.1 | 8 | 1360 | |||
XM_008766842.1 | XP_008765064.1 | 6 | 1254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
