siRNA Details
Gene Symbol: Unc5cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244421.2 | XP_006244483.1 | 9 | 2089 | ||
XM_006244422.2 | XP_006244484.1 | 8 | 1655 | |||
XM_006244423.2 | XP_006244485.1 | 7 | 1817 | |||
XM_008766843.1 | XP_008765065.1 | 7 | 1817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
