siRNA Details
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034136.1 | NP_001029308.1 | 3, 4 | 431 | ||
GenBank mRNA | BC100081.1 | - | 3, 4 | 431 | 7325700 | |
FQ214924.1 | - | 3, 4 | 464 | |||
FQ229593.1 | - | 3, 4 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
