siRNA Details
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007801.2 | NP_001007802.1 | 4 | 505 | ||
GenBank mRNA | AY387096.1 | - | 4 | 400 | ||
BC087102.1 | - | 4 | 505 | 7192818 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145353, BC019384, AK011822, BC011161, AK042644, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
