siRNA Details
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198756.2 | NP_942051.1 | 1 | 104 | ||
GenBank mRNA | BC058507.1 | - | 1 | 108 | 6889538 | |
FQ215521.1 | - | 1 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
