siRNA Details
Gene Symbol: Enpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106892.1 | NP_001100362.1 | 4 | 1491 | ||
XM_006244598.2 | XP_006244660.1 | 5 | 2887 | |||
XM_006244600.2 | XP_006244662.1 | 4 | 2772 | |||
XM_006244601.2 | XP_006244663.1 | 6 | 2536 | |||
XM_006244602.2 | XP_006244664.1 | 4 | 1473 | |||
XM_008766852.1 | XP_008765074.1 | 5 | 1587 | |||
XM_008766853.1 | XP_008765075.1 | 4 | 1381 |
