siRNA Details
Gene Symbol: Mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106903.1 | NP_001100373.1 | 6 | 491 | ||
XM_008766999.1 | XP_008765221.1 | 5 | 450 | |||
GenBank mRNA | BC168851.1 | - | 5 | 386 | 7323830 | |
FQ224516.1 | - | 5 | 383 |
Gene Symbol: NEWGENE_1308196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767056.1 | XP_008765278.1 | 488 | |||
XM_008767057.1 | XP_008765279.1 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
