siRNA Details
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106905.2 | NP_001100375.1 | 10 | 1597 | ||
XM_006244786.2 | XP_006244848.1 | 10 | 2083 | |||
XM_008767018.1 | XP_008765240.1 | 9 | 1642 |
