siRNA Details
Gene Symbol: Tgfbrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106907.1 | NP_001100377.1 | 3 | 1029 | ||
XM_006244788.2 | XP_006244850.1 | 2 | 627 | |||
XM_006244789.2 | XP_006244851.1 | 2 | 580 | |||
XM_006244790.2 | XP_006244852.1 | 2 | 586 | |||
XM_006244791.2 | XP_006244853.1 | 2 | 606 | |||
XM_006244792.2 | XP_006244854.1 | 2 | 524 | |||
XM_006244793.2 | XP_006244855.1 | 2 | 1349 | |||
XM_008767021.1 | XP_008765243.1 | 2 | 665 |
