siRNA Details
Gene Symbol: RGD1310553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008517.1 | NP_001008517.1 | 3 | 490 | ||
GenBank mRNA | BC086514.1 | - | 3 | 490 | 7134744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
