siRNA Details
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009671.1 | NP_001009671.1 | 2 | 242 | ||
XM_008767080.1 | XP_008765302.1 | 2 | 543 | |||
GenBank mRNA | BC088273.1 | - | 2 | 242 | 7380093 | |
FQ219530.1 | - | 1 | 101 |
