siRNA Details
Gene Symbol: Ppil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175707.3 | NP_783638.1 | 6, 7 | 672 | ||
XM_006244964.2 | XP_006245026.1 | 6, 7 | 742 | |||
GenBank mRNA | AF315802.1 | - | 6, 7 | 460 | ||
BC087645.1 | - | 6, 7 | 672 | 7311008 | ||
FQ224392.1 | - | 6, 7 | 469 |
