siRNA Details
Gene Symbol: Crygc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081660.1 | NP_001075129.1 | 3 | 305 | ||
NM_033483.1 | NP_264077.1 | 2 | 288 | |||
GenBank mRNA | BF523642.1 | - | 2 | 294 | ||
J00717.1 | - | 2 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
