siRNA Details
Gene Symbol: Aamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106920.1 | NP_001100390.1 | 4 | 1446 | ||
XM_006245167.2 | XP_006245229.1 | 4 | 459 | |||
XM_006245168.2 | XP_006245230.1 | 4 | 625 | |||
XM_006245169.2 | XP_006245231.1 | 4 | 622 | |||
GenBank mRNA | BC159424.1 | - | 4 | 410 |
