siRNA Details
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024879.1 | NP_001020050.1 | 6, 7 | 684 | ||
XM_006245184.2 | XP_006245246.1 | 6, 7 | 682 | |||
XM_006245185.2 | XP_006245247.1 | 6, 7 | 783 | |||
XM_006245186.2 | XP_006245248.1 | 6, 7 | 735 | |||
GenBank mRNA | BC097388.1 | - | 6, 7 | 684 | 7453069 | |
FQ229070.1 | - | 3, 4 | 653 | |||
FQ234491.1 | - | 6, 7 | 591 |
