siRNA Details
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106922.2 | NP_001100392.2 | 6 | 839 | ||
XM_006245187.2 | XP_006245249.1 | 6 | 1200 | |||
XM_006245188.2 | XP_006245250.1 | 6 | 1200 | |||
GenBank mRNA | BC168233.1 | - | 6 | 839 | 7442632 |
