siRNA Details
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004252.1 | NP_001004252.1 | 5 | 387 | ||
XM_006245189.2 | XP_006245251.1 | 5 | 423 | |||
GenBank mRNA | BC079364.1 | - | 5 | 387 | 7109296 |

More Information
Other Species Targeted

Gene Ontology Categories:
