siRNA Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106924.1 | NP_001100394.1 | 7 | 1102 | ||
XM_006245373.2 | XP_006245435.1 | 8 | 1251 | |||
XM_006245374.2 | XP_006245436.1 | 8 | 1394 | |||
XM_006245375.2 | XP_006245437.1 | 8 | 1153 | |||
GenBank mRNA | FQ224987.1 | - | 1 | 78 | ||
FQ225203.1 | - | 1 | 31 | |||
FQ225560.1 | - | 1 | 78 | |||
FQ226825.1 | - | 3 | 244 | |||
FQ227560.1 | - | 1 | 90 | |||
FQ232493.1 | - | 1 | 90 | |||
FQ232683.1 | - | 4 | 418 | |||
FQ233663.1 | - | 3 | 243 | |||
FQ234937.1 | - | 2 | 175 | |||
FQ235215.1 | - | 1 | 50 |
