siRNA Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106924.1 | NP_001100394.1 | 7 | 1004 | ||
XM_006245373.2 | XP_006245435.1 | 8 | 1153 | |||
XM_006245374.2 | XP_006245436.1 | 8 | 1296 | |||
XM_006245375.2 | XP_006245437.1 | 8 | 1055 | |||
GenBank mRNA | FQ225617.1 | - | 4 | 320 | ||
FQ226825.1 | - | 3 | 146 | |||
FQ232683.1 | - | 4 | 320 | |||
FQ234937.1 | - | 2 | 77 |
