siRNA Details
Gene Symbol: Ugt1a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175846.2 | NP_787040.2 | 1 | 214 | ||
GenBank mRNA | AF461735.1 | - | 1 | 217 | ||
AY435135.1 | - | 1 | 214 |
Gene Symbol: LOC102553187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767335.1 | XP_008765557.1 | 2 | 3053 | ||
XM_008767336.1 | XP_008765558.1 | 2 | 2952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
