siRNA Details
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009825.1 | NP_001009825.1 | 5 | 326 | ||
XM_008767362.1 | XP_008765584.1 | 5 | 1272 | |||
GenBank mRNA | BC088868.1 | - | 5 | 326 | 7321115 | |
FQ233576.1 | - | 340 |
