siRNA Details
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239736.1 | XP_006239798.1 | 6 | 827 | ||
XM_006239738.1 | XP_006239800.1 | 6 | 859 | |||
XM_237502.7 | XP_237502.5 | 6 | 977 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239736.1 | XP_006239798.1 | 6 | 827 | ||
XM_006239738.1 | XP_006239800.1 | 6 | 859 | |||
XM_237502.7 | XP_237502.5 | 6 | 977 |