siRNA Details
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191815.1 | NP_001178744.1 | 17 | 2045 | ||
XM_006245626.2 | XP_006245688.1 | 17 | 2772 | |||
XM_006245627.2 | XP_006245689.1 | 17 | 2442 | |||
XM_006245628.2 | XP_006245690.2 | 17 | 2530 | |||
XM_008767424.1 | XP_008765646.1 | 17 | 1952 | |||
XM_008767425.1 | XP_008765647.1 | 17 | 2287 |
