siRNA Details
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106928.1 | NP_001100398.1 | 5 | 705 | ||
XM_006245600.2 | XP_006245662.1 | 10 | 1434 | |||
XM_008767367.1 | XP_008765589.1 | 10 | 2210 | |||
XM_008767369.1 | XP_008765591.1 | 11 | 1572 | |||
XM_008767370.1 | XP_008765592.1 | 10 | 2470 | |||
XM_008767371.1 | XP_008765593.1 | 10 | 2210 | |||
XM_008767372.1 | XP_008765594.1 | 3 | 350 |
