siRNA Details
Gene Symbol: Ssty1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_229292.8 | XP_229292.6 | 1 | 493 |
Gene Symbol: LOC102553316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235107.2 | XP_006235169.2 | 1 | 493 |
Gene Symbol: LOC103694737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773755.1 | XP_008771977.1 | 1 | 505 |
