siRNA Details
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106930.1 | NP_001100400.1 | 4 | 567 | ||
XM_008768871.1 | XP_008767093.1 | 4 | 598 | |||
GenBank mRNA | FQ216783.1 | - | 3 | 443 |
