siRNA Details
Gene Symbol: LOC689479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750610.3 | XP_003750658.2 | 5 | 1393 | ||
XM_008766735.1 | XP_008764957.1 | 4 | 1064 |
Gene Symbol: LOC302192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253274.2 | XP_006253336.1 | 1053 | |||
XM_006253276.2 | XP_006253338.1 | 1010 | ||||
XM_217539.8 | XP_217539.4 | 779 | ||||
GenBank mRNA | BC082094.1 | - | 988 | |||
BC100254.1 | - | 990 | 7456988 |
Gene Symbol: LOC681180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253263.2 | XP_006253325.1 | 5 | 1010 | ||
XM_008771334.1 | XP_008769556.1 | 4 | 1062 |
