siRNA Details
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106937.1 | NP_001100407.1 | 8 | 942 | ||
XM_006257159.2 | XP_006257221.1 | 9 | 1211 | |||
XM_006257160.2 | XP_006257222.1 | 9 | 1061 | |||
XM_006257161.2 | XP_006257223.1 | 9 | 1208 | |||
XM_006257162.2 | XP_006257224.1 | 8 | 1251 | |||
XM_008773339.1 | XP_008771561.1 | 8 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
