siRNA Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257110.2 | XP_006257172.1 | 7 | 1105 | ||
XM_006257111.2 | XP_006257173.1 | 7 | 1105 | |||
XM_006257112.2 | XP_006257174.1 | 7 | 1096 | |||
XM_228582.8 | XP_228582.6 | 7 | 1096 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177938, NM_001177937, NM_001177944, NM_001177939, NM_001177943, and others.
Gene Ontology Categories:
