siRNA Details
Gene Symbol: RGD1563590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257014.2 | XP_006257076.2 | 1 | 1014 |
Gene Symbol: RGD1559970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773314.1 | XP_008771536.1 | 1 | 568 |
