siRNA Details
Gene Symbol: LOC100909486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773311.1 | XP_008771533.1 | 1 | 473 |
Gene Symbol: RGD1562980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773312.1 | XP_008771534.1 | 1 | 428 |
