siRNA Details
Gene Symbol: Slitrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106947.1 | NP_001100417.1 | 3 | 803 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178740, AK053065, AK039763, BC117891, AK046301, and others.
Gene Ontology Categories:
