siRNA Details
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106948.1 | NP_001100418.1 | 3 | 2849 | ||
XM_008773555.1 | XP_008771777.1 | 2 | 2693 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106948.1 | NP_001100418.1 | 3 | 2849 | ||
XM_008773555.1 | XP_008771777.1 | 2 | 2693 |