siRNA Details
Gene Symbol: Mbnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106949.1 | NP_001100419.1 | 3 | 465 | ||
XM_008773556.1 | XP_008771778.1 | 3 | 1944 | |||
XM_008773557.1 | XP_008771779.1 | 3 | 1067 | |||
XM_008773558.1 | XP_008771780.1 | 3 | 1064 | |||
XM_008773559.1 | XP_008771781.1 | 3 | 1067 | |||
XM_008773560.1 | XP_008771782.1 | 3 | 1070 | |||
XM_008773561.1 | XP_008771783.1 | 3 | 1067 | |||
XM_008773562.1 | XP_008771784.1 | 3 | 340 |
