siRNA Details
Gene Symbol: Mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044237.1 | NP_001037702.1 | 3 | 798 | ||
XM_006257488.2 | XP_006257550.1 | 3 | 627 | |||
GenBank mRNA | BC098848.1 | - | 3 | 798 | 7379624 | |
FQ211690.1 | - | 2 | 233 |
