siRNA Details
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178333.1 | NP_848016.1 | 2, 3 | 336 | ||
XM_008773069.1 | XP_008771291.1 | 4, 5 | 1666 | |||
XM_008773070.1 | XP_008771292.1 | 4, 5 | 1666 | |||
GenBank mRNA | AB098162.1 | - | 2, 3 | 336 |
