siRNA Details
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106956.1 | NP_001100426.1 | 3 | 1240 | ||
XM_006256703.2 | XP_006256765.1 | 3 | 1456 | |||
XM_006256704.2 | XP_006256766.1 | 3 | 1032 | |||
XM_006256705.2 | XP_006256767.1 | 3 | 1033 | |||
GenBank mRNA | BC160871.1 | - | 3 | 807 | 7364965 |
