siRNA Details
Gene Symbol: LOC302576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013959.1 | NP_001013981.1 | 1 | 1394 | ||
GenBank mRNA | BC083583.1 | - | 1 | 1394 | 7131068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
