siRNA Details
Gene Symbol: Tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191618.2 | NP_001178547.1 | 6 | 1762 | ||
XM_006257038.2 | XP_006257100.1 | 6 | 1916 |

More Information
Other Species Targeted

Gene Ontology Categories:
