siRNA Details
Gene Symbol: Gpr143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106958.1 | NP_001100428.1 | 7 | 868 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106958.1 | NP_001100428.1 | 7 | 868 |