siRNA Details
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007667.2 | NP_001007668.1 | 4 | 557 | ||
XM_008773228.1 | XP_008771450.1 | 4 | 1557 | |||
XM_008773229.1 | XP_008771451.1 | 4 | 953 | |||
XM_008773230.1 | XP_008771452.1 | 5 | 592 | |||
GenBank mRNA | AY325208.1 | - | 422 | |||
BC060588.1 | - | 4 | 559 | 6921572 | ||
FQ222039.1 | - | 4 | 439 | |||
FQ229984.1 | - | 4 | 441 |
