siRNA Details
Gene Symbol: Ofd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106961.1 | NP_001100431.1 | 11 | 1473 | ||
XM_008773162.1 | XP_008771384.1 | 11 | 1436 | |||
XM_008773163.1 | XP_008771385.1 | 11 | 1435 | |||
XM_008773164.1 | XP_008771386.1 | 11 | 1129 | |||
XM_008773165.1 | XP_008771387.1 | 10 | 1235 | |||
GenBank mRNA | FQ210045.1 | - | 10 | 996 |
