siRNA Details
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256858.2 | XP_006256920.1 | 1 | 410 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK146664, AK005404, AK002412, AF398972, AK078851.
Gene Ontology Categories:
