siRNA Details
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256887.2 | XP_006256949.1 | 8, 9 | 807 | ||
XM_217612.5 | XP_217612.5 | 7, 8 | 930 |

More Information
Other Species Targeted

Gene Ontology Categories:
